Merck
  • Intercellular transfer of messenger RNAs in multiorgan tumorigenesis by tumor cell-derived exosomes.
Intercellular transfer of messenger RNAs in multiorgan tumorigenesis by tumor cell-derived exosomes.
Molecular medicine reports (2015-02-13)
Hong Jiang, Zheng Li, Xiaohua Li, Jianguo Xia
ABSTRACT
Exosomes are small membrane vesicles of endocytic origin. They are derived from various cells, including tumor cells, and may serve as important modulators of intercellular communication. The present study established a U‑87 MG human glioblastoma cell line that showed a stable expression of green fluorescent protein (GFP; U‑87‑GFP). Two types of human tumor cell lines, U‑87‑GFP and LoVo human colon cancer cells, were demonstrated to communicate through the transfer of GFP messenger (m)RNA by exosomes. Furthermore, GFP mRNAs delivered by the U‑87‑GFP cell‑derived exosomes were translated into functional proteins in the recipient LoVo cells. In addition, LoVo cells were demonstrated to uptake exosomes derived from the U‑87‑GFP cells by clathrin‑mediated endocytosis. These results indicate that tumor cell‑derived exosomes may represent vesicular carriers that regulate gene expression, which may provide a pathway of intercellular communication in the tumor microenvironment during multiorgan tumorigenesis. The exosome uptake pathway may have potential therapeutic applications in multiorgan tumorigenesis.
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications