Merck
  • Comparative investigations for adenovirus recognition and quantification: Plastic or natural antibodies?
Comparative investigations for adenovirus recognition and quantification: Plastic or natural antibodies?
Biosensors & bioelectronics (2015-08-13)
Zeynep Altintas, Jack Pocock, Katy-Anne Thompson, Ibtisam E Tothill
ABSTRACT
Comparative and comprehensive investigations for adenovirus recognition and detection were conducted using plastic and natural antibodies to compare three different strategies. The implementation of molecularly imprinted polymer (MIP) technology for specific and sensitive recognition of viruses with the combination of biosensors was reported. Plastic antibodies (MIPs nanoparticles) were produced for adenovirus by employing a novel solid phase synthesis method. MIP receptors were then characterised using dynamic light scattering (DLS) and transmission electron microscopy (TEM) techniques prior to immobilisation on a surface plasmon resonance (SPR) sensor as affinity receptor for adenovirus detection. Two different templates were also imprinted as control MIPs (vancomycin-MIP and MS2-MIP). The specific recognition of adenovirus was investigated in the concentration range of 0.01-20 pM and the limit of detection was achieved as 0.02 pM. As an alternative to MIP receptors, direct and sandwich assays were developed for adenovirus quantification using natural antibodies. The detection limit of direct and sandwich assays were found as 0.3 pM and 0.008 pM, respectively. The kinetic data analyses were performed for three different adenovirus recognition methods and cross-reactivity studies were also conducted using MS2 phage as control virus and an excellent specificity was achieved with all assays types. This work confirmed the suitability of the MIPs SPR sensor for the detection of viruses.
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)