Merck
  • Natural product extracts of the Canadian prairie plant, Thermopsis rhombifolia, have anti-cancer activity in phenotypic cell-based assays.
Natural product extracts of the Canadian prairie plant, Thermopsis rhombifolia, have anti-cancer activity in phenotypic cell-based assays.
Natural product research (2014-11-26)
Sophie Kernéis, Lucy H Swift, Cody W Lewis, Céline Bruyère, Nassima Oumata, Pierre Colas, Sandrine Ruchaud, John Bain, Roy M Golsteyn
ABSTRACT
Many plant species within the terrestrial ecological zones of Canada have not yet been investigated for anti-cancer activity. We examined the scientific literature describing the endemic flora from the prairie ecological zone and selected the species, Thermopsis rhombifolia, locally known as the buffalo bean, for investigation of its anti-cancer potential. We tested it in cell-based assays using phenotypic screens that feature some of the hallmarks of cancer. An ethanolic extract prepared from T. rhombifolia was cytotoxic to HT-29 (colon) and SH-SY5Y (brain) cancer cell lines, and showed little cytotoxicity to a normal human cell line (WI-38). In phenotypic assays, we identified activities in the extracts that target cell death, cell cycle and cell adhesion. These data highlight the anti-cancer potential of previously untested plants found in northern ecological zones and the feasibility of using pertinent phenotypic assays to examine the anti-cancer potential of natural product extracts.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
CR8, ≥95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis