Merck
  • Three Series of Sulfo-Functionalized Mixed-Linker CAU-10 Analogues: Sorption Properties, Proton Conductivity, and Catalytic Activity.
Three Series of Sulfo-Functionalized Mixed-Linker CAU-10 Analogues: Sorption Properties, Proton Conductivity, and Catalytic Activity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-30)
Nele Reimer, Bart Bueken, Sebastian Leubner, Christopher Seidler, Michael Wark, Dirk De Vos, Norbert Stock
ABSTRACT
Ten mixed-linker metal-organic frameworks [Al(OH)(m-BDC-X)(1-y)(m-BDC-SO3H)y] (H2BDC = 1,3-benzenedicarboxylic acid; X = H, NO2, OH) exhibiting the CAU-10-type structure were synthesized. The compounds can be grouped into three series according to the combination of ligands employed. The three series of compounds were obtained by employing different ratios of m-H2 BDC-X and m-H2BDC-SO3Li. The resulting compounds, which are denoted CAU-10-H/Sx, -N/Sx and -O/Sx, show exceptionally high thermal stability for sulfonated materials of up to 350 °C. Detailed characterization with special focus on polarity and acidity was performed, and the impact of the additional SO3H groups is clearly demonstrated by changes in the sorption affinities/capacities towards several gases and water vapor. In addition, selected samples were evaluated for proton conductivity and as catalysts for the gas-phase dehydration of ethanol to ethylene. While only very low proton conductivities were observed, a pronounced increase in catalytic activity was achieved. Although reactions were performed at temperatures of 250 and 300 °C for more than 40 h, no desulfonation and no loss of crystallinity were observed, and stable ethanol conversion resulted. This demonstrates the high stability of this material.
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis