Merck
  • Identification of Sex-Linked SNPs and Sex-Determining Regions in the Yellowtail Genome.
Identification of Sex-Linked SNPs and Sex-Determining Regions in the Yellowtail Genome.
Marine biotechnology (New York, N.Y.) (2015-05-16)
Takashi Koyama, Akiyuki Ozaki, Kazunori Yoshida, Junpei Suzuki, Kanako Fuji, Jun-ya Aoki, Wataru Kai, Yumi Kawabata, Tatsuo Tsuzaki, Kazuo Araki, Takashi Sakamoto
ABSTRACT
Unlike the conservation of sex-determining (SD) modes seen in most mammals and birds, teleost fishes exhibit a wide variety of SD systems and genes. Hence, the study of SD genes and sex chromosome turnover in fish is one of the most interesting topics in evolutionary biology. To increase resolution of the SD gene evolutionary trajectory in fish, identification of the SD gene in more fish species is necessary. In this study, we focused on the yellowtail, a species widely cultivated in Japan. It is a member of family Carangidae in which no heteromorphic sex chromosome has been observed, and no SD gene has been identified to date. By performing linkage analysis and BAC walking, we identified a genomic region and SNPs with complete linkage to yellowtail sex. Comparative genome analysis revealed the yellowtail SD region ancestral chromosome structure as medaka-fugu. Two inversions occurred in the yellowtail linage after it diverged from the yellowtail-medaka ancestor. An association study using wild yellowtails and the SNPs developed from BAC ends identified two SNPs that can reasonably distinguish the sexes. Therefore, these will be useful genetic markers for yellowtail breeding. Based on a comparative study, it was suggested that a PDZ domain containing the GIPC protein might be involved in yellowtail sex determination. The homomorphic sex chromosomes widely observed in the Carangidae suggest that this family could be a suitable marine fish model to investigate the early stages of sex chromosome evolution, for which our results provide a good starting point.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis
Sigma-Aldrich
BES, ≥99.0% (titration)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O