Merck
  • CD155 (PVR/Necl5) mediates a costimulatory signal in CD4+ T cells and regulates allergic inflammation.
CD155 (PVR/Necl5) mediates a costimulatory signal in CD4+ T cells and regulates allergic inflammation.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-15)
Yumi Yamashita-Kanemaru, Yuichi Takahashi, Yinan Wang, Satoko Tahara-Hanaoka, Shin-Ichiro Honda, Günter Bernhardt, Akira Shibuya, Kazuko Shibuya
ABSTRACT
Although Th1 and Th2 cells are known to be involved in allergic inflammatory diseases, the molecular mechanisms underlying their differentiation are incompletely understood. In this study, we identified CD155 as a costimulatory molecule on CD4(+) T cells. Importantly, CD155-mediated signaling induced Th1 development in both humans and mice, as evidenced by production of IFN-γ and upregulation of Tbx21 transcription; these effects were independent of IL-12 but dependent on NF-κB-induced autocrine IFN-γ that triggered positive feedback via STAT1 activation. Mice genetically deficient in CD155 or treated with anti-CD155 Ab exhibited attenuated Th1-type contact hypersensitivity. Thus, CD155 plays an important regulatory role in helper T cell differentiation and allergic diseases.
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%