Merck
  • Development of an LC-MS/MS method for studying migration characteristics of acetaldehyde in polyethylene terephthalate (PET)-packed mineral water.
Development of an LC-MS/MS method for studying migration characteristics of acetaldehyde in polyethylene terephthalate (PET)-packed mineral water.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-08-11)
Nina Baumjohann, Diedrich Harms
ABSTRACT
During storage, acetaldehyde migration from polyethylene terephthalate (PET) bottles can affect the quality of mineral water even in the low µg l(-1) range negatively, as it features a fruity or plastic-like off-flavour. For a sensitive and fast analysis of acetaldehyde in mineral water, a new analysis method of 2,4-dinitrophenylhydrazine (DNPH) derivatisation followed by HPLC-electrospray tandem mass spectrometry (ESI-MS/MS) was developed. Acetaldehyde was directly derivatised in the mineral water sample avoiding extraction and/or pre-concentration steps and then analysed by reversed-phase HPLC-ESI-MS/MS using multiple reaction monitoring mode (MRM). Along with method development, the optimum molar excess of DNPH in contrast to acetaldehyde was studied for the mineral water matrix, because no specific and robust data were yet available for this critical parameter. Best results were obtained by using a calibration via the derivatisation reaction. Without any analyte enrichment or extraction, an LOD of 0.5 µg l(-1) and an LOQ of 1.9 µg l(-1) were achieved. Using the developed method, mineral water samples packed in PET bottles from Germany were analysed and the correlation between the acetaldehyde concentration and other characteristics of the samples was evaluated illustrating the applicability of the method. Besides a relationship between bottle size and CO2 content of the mineral water and acetaldehyde migration, a correlation with acetaldehyde migration and the material composition of the bottle, e.g. recycled PET, was noted. Investigating the light influence on the acetaldehyde migration with a newly developed, reproducible light exposure setup, a significant increase of the acetaldehyde concentration in carbonated mineral water samples was observed.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC