Merck
  • The species-specific regenerative effects of notochordal cell-conditioned medium on chondrocyte-like cells derived from degenerated human intervertebral discs.
The species-specific regenerative effects of notochordal cell-conditioned medium on chondrocyte-like cells derived from degenerated human intervertebral discs.
European cells & materials (2015-09-22)
F C Bach, S A H de Vries, A Krouwels, L B Creemers, K Ito, B P Meij, M A Tryfonidou
ABSTRACT
During intervertebral disc (IVD) maturation, the main cell type shifts from notochordal cells (NCs) to chondrocyte-like cells (CLCs). NCs secrete factors with regenerative potential, making them an interesting focus for regenerative treatments. During initial development, these strategies preferably employ non-human donors due to easy availability of their NC-rich nucleus pulposus (NP) tissue. To increase the success of translating these strategies for clinical application, this study aimed to delineate whether NC-secreted factors of different species have a regenerative effect on human CLCs. Human, canine and porcine NC-rich NP tissue and NC-conditioned medium (NCCM) were analysed biochemically and histologically. Human CLC micro-aggregates from degenerated IVDs were cultured in human, canine or porcine NCCM. Collagen, glycosaminoglycan (GAG) and DNA content was determined and histology was performed. Canine and porcine NPs were richer in NCs than human NPs. Human NPs contained the highest collagen content, whereas the DNA and GAG content of canine NPs was significantly higher than that of human or porcine NPs. NCCM from all species significantly increased the DNA and GAG content of the human CLC micro-aggregates. Porcine and canine NCCM were significantly more potent than human NCCM in inducing GAG deposition, whereas only human NCCM induced collagen type II production. Secreted factors from human, canine and porcine NC-rich NPs exerted regenerative effects on human CLCs, indicating a cross-species effect. Bioactive compound(s) are present in NCCM of different species that may reverse human IVD degeneration, supporting further research into strategies based on NC-technology employing canine or porcine models for their translation into humans.
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
SAFC
L-Proline
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)