Merck
  • Quantitative fingerprinting of O-linked glycans released from proteins using isotopic coded labeling with deuterated 1-phenyl-3-methyl-5-pyrazolone.
Quantitative fingerprinting of O-linked glycans released from proteins using isotopic coded labeling with deuterated 1-phenyl-3-methyl-5-pyrazolone.
Journal of chromatography. A (2015-07-18)
Siniša Sić, Norbert M Maier, Andreas M Rizzi
ABSTRACT
Investigation of oligosaccharides attached to proteins as post-translational modification remains an important research field in the area of glycoproteomics as well as in biotechnology. The development of new tools for qualitative and quantitative analysis of glycans has gained high importance in recent years. This is particularly true with O-glycans for which quantitative data are still underrepresented in literature. This fact is probably due to the absence of an enzyme for general release of O-linked saccharides from glycoproteins and due to their low ionization yield in mass spectrometry (MS). In this paper, a method is established aimed at improved qualitative and quantitative analysis of mucin-type O-glycans. A chemical reaction combining release and derivatization of O-glycans in one step is combined here with mass spectrometric quantification. For the purpose of improved quantitative analysis, stable-isotope coded labeling by d0/d5 1-phenyl-3-methyl-5-pyrazolidone (PMP) was performed. The "heavy"-version of this label, penta-deutero (d5)-PMP, was synthesized for this purpose. Beneath improving the reproducibility of quantitation, PMP derivatization contributed to an enhancement of ionization yields in MS. By introducing an internal standard (e.g. GlcNAc3) the reproducibility for quantification can be improved. For higher abundant O-glycans a mean coefficient of variation (CV) less than 6% could be attained, for very low abundant CV values between 15 and 20%. For the determination of O-glycan profiles in mixtures, a HPLC separation was combined with a high resolution Qq-oaTOF instrument. RP-type stationary phases were successful in separating glycan species including some of isomeric ones. This separation step was particularly useful for removing of salts avoiding so the presence of various sodium clusters in the MS spectrum.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)