Merck
  • Ethanol Extract of Sarcodon asparatus Mitigates Inflammatory Responses in Lipopolysaccharide-Challenged Mice and Murine Macrophages.
Ethanol Extract of Sarcodon asparatus Mitigates Inflammatory Responses in Lipopolysaccharide-Challenged Mice and Murine Macrophages.
Journal of medicinal food (2015-04-04)
Min-Yu Chung, Sung Keun Jung, Hye-Jin Lee, Dong Hwa Shon, Hyun-Ku Kim
ABSTRACT
A number of compounds isolated from mushrooms have exhibited disease-modifying effects. We sought to investigate the mechanisms responsible for the anti-inflammatory effects of an extract from the mushroom species Sarcodon asparatus (SAE). Male BALB/c mice (N=42; 6 weeks old) were randomly assigned to four treatment groups. Intraperitoneal administration of SAE significantly attenuated lipopolysaccharide (LPS)-mediated increases in alanine aminotransferase (ALT) activity. LPS also increased serum levels of proinflammatory cytokines, including IL-1β, IL-6, and TNF-α, which were dose-dependently and significantly attenuated by SAE. Correlative relationships between serum ALT activity and proinflammatory cytokines suggested that SAE-mediated suppression of liver injury was partly attributable to the attenuation of serum inflammatory responses. SAE significantly decreased hepatic NO(•) production and subsequent 3-nitrotyrosine formation, and the hepatic NO(•) production significantly correlated with serum ALT and cytokine levels, suggesting that SAE mitigates liver injury in association with inflammatory processes, likely by suppressing NO(•) production. Anti-inflammatory activity and further mechanisms of SAE were evaluated using RAW264.7 with LPS challenge. Noncytotoxic levels of SAE significantly attenuated NO(•) production in RAW264.7 cells and also markedly suppressed the expression of iNOS and other proinflammatory mediators, including COX-2 and IL-6, which were upregulated in the presence of LPS. SAE inhibited the phosphorylation of p65, an observation that occurred independently of IKKαβ-mediated IκBα phosphorylation. Collectively, our results demonstrate that SAE suppressed NO(•)-mediated inflammation by inhibiting p65 transcriptional activation without affecting IKKαβ-mediated IκBα phosphorylation. Further studies are warranted to examine the major compounds responsible for these effects and the mechanisms responsible for the p65 phosphorylation observed.
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology