Merck
  • Duration of Colonization and Determinants of Earlier Clearance of Colonization With Methicillin-Resistant Staphylococcus aureus.
Duration of Colonization and Determinants of Earlier Clearance of Colonization With Methicillin-Resistant Staphylococcus aureus.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2015-02-05)
Valerie C Cluzet, Jeffrey S Gerber, Irving Nachamkin, Joshua P Metlay, Theoklis E Zaoutis, Meghan F Davis, Kathleen G Julian, David Royer, Darren R Linkin, Susan E Coffin, David J Margolis, Judd E Hollander, Rakesh D Mistry, Laurence J Gavin, Pam Tolomeo, Jacqueleen A Wise, Mary K Wheeler, Warren B Bilker, Xiaoyan Han, Baofeng Hu, Neil O Fishman, Ebbing Lautenbach
ABSTRACT
The duration of colonization and factors associated with clearance of methicillin-resistant Staphylococcus aureus (MRSA) after community-onset MRSA skin and soft-tissue infection (SSTI) remain unclear. We conducted a prospective cohort study of patients with acute MRSA SSTI presenting to 5 adult and pediatric academic hospitals from 1 January 2010 through 31 December 2012. Index patients and household members performed self-sampling for MRSA colonization every 2 weeks for 6 months. Clearance of colonization was defined as negative MRSA surveillance cultures during 2 consecutive sampling periods. A Cox proportional hazards regression model was developed to identify determinants of clearance of colonization. Two hundred forty-three index patients were included. The median duration of MRSA colonization after SSTI diagnosis was 21 days (95% confidence interval [CI], 19-24), and 19.8% never cleared colonization. Treatment of the SSTI with clindamycin was associated with earlier clearance (hazard ratio [HR], 1.72; 95% CI, 1.28-2.30; P < .001). Older age (HR, 0.99; 95% CI, .98-1.00; P = .01) was associated with longer duration of colonization. There was a borderline significant association between increased number of household members colonized with MRSA and later clearance of colonization in the index patient (HR, 0.85; 95% CI, .71-1.01; P = .06). With a systematic, regular sampling protocol, duration of MRSA colonization was noted to be shorter than previously reported, although 19.8% of patients remained colonized at 6 months. The association between clindamycin and shorter duration of colonization after MRSA SSTI suggests a possible role for the antibiotic selected for treatment of MRSA infection.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)