Merck
  • Rapid, sensitive and selective HPLC-MS/MS method for the quantification of topically applied besifloxacin in rabbit plasma and ocular tissues: Application to a pharmacokinetic study.
Rapid, sensitive and selective HPLC-MS/MS method for the quantification of topically applied besifloxacin in rabbit plasma and ocular tissues: Application to a pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2015-09-05)
Xiao-Fei Gu, Bai-Yang Mao, Min Xia, Yang Yang, Jia-Li Zhang, Da-Song Yang, Wei-Xin Wu, Ying-Xiang Du, Bin Di, Meng-Xiang Su
ABSTRACT
Besifloxacin is a fourth-generation broad-spectrum fluoroquinolone registered for the topical treatment of bacterial conjunctivitis. In this study, a rapid, sensitive and selective liquid chromatography tandem mass spectrometry (LC-MS/MS) method was developed for quantification of besifloxacin in rabbit plasma and ocular tissues using nateglinide as the internal standard (IS). The analyte and IS were separated on a Sepax GP-Phenyl column by isocratic elution with methanol-acetonitrile-5 mM ammonium formate-formic acid (29:55:16:0.1, v/v/v/v) as the mobile phase at a flow rate of 1.2 mL/min, and the total run time was 3.0 min. An electrospray ionization (ESI) source was applied and operated in the positive ion mode; multiple reaction monitoring (MRM) mode was used for quantification, and the monitored transitions were 394.2→377.1 for besifloxacin and m/z 318.3→166.1 for the IS. The calibration curve was linear over the range of 0.103-206 ng/mL for plasma and 2.06-2060 ng/mL for tears, aqueous humor, conjunctiva and cornea with correlation coefficient (r) greater than 0.99. The lower limit of quantification (LLOQ) for besifloxacin was 0.103 ng/mL for plasma and 2.06 ng/mL for other ocular tissues with good accuracy and precision. Intra- and inter-batch precision were both lower than 15% and accuracy ranged from 85% to 115% at all QC levels. The method was successfully applied to the pharmacokinetic study of besifloxacin in rabbit plasma and ocular tissues after single and multiple topical administrations.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%