Merck
  • Recombination between Streptococcus suis ICESsu32457 and Streptococcus agalactiae ICESa2603 yields a hybrid ICE transferable to Streptococcus pyogenes.
Recombination between Streptococcus suis ICESsu32457 and Streptococcus agalactiae ICESa2603 yields a hybrid ICE transferable to Streptococcus pyogenes.
Veterinary microbiology (2015-05-04)
Emanuela Marini, Claudio Palmieri, Gloria Magi, Bruna Facinelli
ABSTRACT
Integrative conjugative elements (ICEs) are mobile genetic elements that reside in the chromosome but retain the ability to undergo excision and to transfer by conjugation. Genes involved in drug resistance, virulence, or niche adaptation are often found among backbone genes as cargo DNA. We recently characterized in Streptococcus suis an ICE (ICESsu32457) carrying resistance genes [tet(O/W/32/O), tet(40), erm(B), aphA, and aadE] in the 15K unstable genetic element, which is flanked by two ∼1.3kb direct repeats. Remarkably, ∼1.3-kb sequences are conserved in ICESa2603 of Streptococcus agalactiae 2603V/R, which carry heavy metal resistance genes cadC/cadA and mer. In matings between S. suis 32457 (donor) and S. agalactiae 2603V/R (recipient), transconjugants were obtained. PCR experiments, PFGE, and sequence analysis of transconjugants demonstrated a tandem array between ICESsu32457 and ICESa2603. Matings between tandem array-containing S. agalactiae 2603V/R (donor) and Streptococcus pyogenes RF12 (recipient) yielded a single transconjugant containing a hybrid ICE, here named ICESa2603/ICESsu32457. The hybrid formed by recombination of the left ∼1.3-kb sequence of ICESsu32457 and the ∼1.3-kb sequence of ICESa2603. Interestingly, the hybrid ICE was transferable between S. pyogenes strains, thus demonstrating that it behaves as a conventional ICE. These findings suggest that both tandem arrays and hybrid ICEs may contribute to the evolution of antibiotic resistance in streptococci, creating novel mobile elements capable of disseminating new combinations of antibiotic resistance genes.
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Fusidic acid
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications