Merck
  • CD44 Influences Fibroblast Behaviors Via Modulation of Cell-Cell and Cell-Matrix Interactions, Affecting Survivin and Hippo Pathways.
CD44 Influences Fibroblast Behaviors Via Modulation of Cell-Cell and Cell-Matrix Interactions, Affecting Survivin and Hippo Pathways.
Journal of cellular physiology (2015-08-08)
Masayuki Tsuneki, Joseph A Madri
ABSTRACT
CD44 has been studied in a wide variety of cell types, in a diverse array of cell behaviors and in a diverse range of signaling pathways. We now document a role for CD44 in mediating fibroblast behaviors via regulation of N-cadherin, extracellular matrix expression, Survivin and the Hippo pathway. Here, we report our findings on the roles of CD44 in modulating proliferation, apoptosis, migration and invasion of murine wild-type (WT-FB) and CD44 knockout dermal fibroblasts (CD44KO-FB). As we have documented in microvascular endothelial cells lacking CD44, we found persistent increased proliferation, reduced activation of cleaved caspase 3, increased initial attachment, but decreased strength of cell attachment in high cell density, post confluent CD44KO-FB cultures. Additionally, we found that siRNA knock-down of CD44 mimicked the behaviors of CD44KO-FB, restoring the decreases in N-cadherin, collagen type I, fibronectin, Survivin, nuclear fractions of YAP and phospho-YAP and decreased levels of cleaved caspase 3 to the levels observed in CD44KO-FB. Interestingly, plating CD44KO-FB on collagen type I or fibronectin resulted in significant decreases in secondary proliferation rates compared to plating cells on non-coated dishes, consistent with increased cell adhesion compared to their effects on WT-FB. Lastly, siRNA knockdown of CD44 in WT-FB resulted in increased fibroblast migration compared to WT-FB, albeit at reduced rates compared to CD44KO-FB. These results are consistent with CD44's pivotal role in modulating several diverse behaviors important for adhesion, proliferation, apoptosis, migration and invasion during development, growth, repair, maintenance and regression of a wide variety of mesenchymal tissues.
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
DAPI, for nucleic acid staining
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting human CD44