Merck
  • Internalization of GluA2 and the underlying mechanisms of cognitive decline in aged rats following surgery and prolonged exposure to sevoflurane.
Internalization of GluA2 and the underlying mechanisms of cognitive decline in aged rats following surgery and prolonged exposure to sevoflurane.
Neurotoxicology (2015-06-06)
Nan Hu, Miaomiao Wang, Keliang Xie, Haiyun Wang, Chenxu Wang, Chao Wang, Chunyan Wang, Yize Li, Yonghao Yu, Guolin Wang
ABSTRACT
We revealed that a high concentration of sevoflurane exacerbated cognitive impairment in aged rats, and the inhibition of GluA2 subunit internalization facilitated neuroprotection after a cerebral ischemic injury. However, the trafficking of GluA2 in POCD and its underlying mechanism are not clear. We thus detected the effects of sevoflurane for different inhalation durations on postoperative cognitive function and investigated the role of GluA2 subunit trafficking in this process. A rat model of orthopedic surgery was performed with different durations of 1.5 MAC sevoflurane inhalation. Cognitive function was evaluated by manipulating the Y maze and fear conditioning tests for 7 days after experiments. Western blot, ELISA and coimmunoprecipitation were applied to analyze GluA2 internalization, PI3K expression and its activity, as well as alterations to the MEF2-Arc pathway in the hippocampus. Neuron apoptosis and the spine morphology in the hippocampus were also observed. We found that neuron apoptosis and GluA2 internalization increased following surgery and 1.5 MAC sevoflurane inhalation for 2h, possibly due to the decrease of the PI3K-GluA2 complex and PI3K activity in the hippocampus after prolonged 1.5 MAC sevoflurane inhalation. We also observed that the MEF2-Arc pathway contributed to long-term cognitive function, which also impaired the spinal morphology after 1.5 MAC sevoflurane inhalation for 2h. The above results suggest that 1.5 MAC sevoflurane inhalation for 2h potentiated surgery-impaired cognitive function and that the inhibition of PI3K-AMPAR GluA2 as well as activation of the MEF2-Arc signal pathway contributes to different stages of POCD.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O