Merck
  • Negligible Pharmacokinetic Interaction of Red Ginseng and Losartan, an Antihypertensive Agent, in Sprague-Dawley Rats.
Negligible Pharmacokinetic Interaction of Red Ginseng and Losartan, an Antihypertensive Agent, in Sprague-Dawley Rats.
Journal of toxicology and environmental health. Part A (2015-10-31)
Sung Ha Ryu, Yong Soon Kim, Hyun-Jun Jang, Kyu-Bong Kim
ABSTRACT
Red ginseng (RG) is one of the top selling herbal medicines in Korea, but is not recommended in hypertensive patients. In this study, the pharmacokinetic (PK) interaction between RG and losartan, an antihypertensive drug, was examined. RG was orally administered for 2 wk to male Sprague-Dawley (S-D) rats at either control (0), 0.5, 1, or 2 g/kg/d for 2 wk. After the last administration of RG and 30 min later, all animals were treated with 10 mg/kg losartan by oral route. In addition, some S-D rats were administered RG orally for 21 d at 2 g/kg followed by losartan intravenously (iv) at 10 mg/kg/d. Post losartan administration, plasma samples were collected at 5, 15, and 30 min and 1, 1.5, 2, 3, 6, 12, and 24 h. Plasma concentrations of losartan and E-3174, the active metabolite of losartan, were analyzed by a high-pressure liquid chromatography-tandem mass spectrometer system (LC-MS/MS). Oral losartan administration showed dose-dependent pharmacokinetics (PK) increase with time to maximum plasma, but this was not significant between different groups. There was no significant change in tmax with E-3174 PK. With iv losartan, pharmacokinetics showed elevation of area under the plasma concentration-time curve from time zero extrapolated to infinitity. There was not a significant change in AUCinf with E-3174 PK. Therefore, RG appeared to interfere with biotransformation of losartan, as RG exerted no marked effect on E-3174 PK in S-D rats. Data demonstrated that oral or iv treatment with losartan in rats pretreated with RG for 2 wk showed that losartan PK was affected but E-3174 PK remained unchanged among different dose groups. These results suggested that RG induces negligible influence on losartan and E-3174 PK in rats.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%