Merck
  • Gold nanoparticles immobilized hydrophilic monoliths with variable functional modification for highly selective enrichment and on-line deglycosylation of glycopeptides.
Gold nanoparticles immobilized hydrophilic monoliths with variable functional modification for highly selective enrichment and on-line deglycosylation of glycopeptides.
Analytica chimica acta (2015-11-18)
Yu Liang, Ci Wu, Qun Zhao, Qi Wu, Bo Jiang, Yejing Weng, Zhen Liang, Lihua Zhang, Yukui Zhang
ABSTRACT
The poly (glycidyl methacrylate-co-poly (ethylene glycol) diacrylate) monoliths modified with gold nanoparticles, with advantages of enhanced reactive sites, good hydrophilicity and facile modification, were prepared as the matrix, followed by variable functionalization with cysteine and PNGase F for glycopeptide enrichment and on-line deglycosylation respectively. By the cysteine functionalized monolithic column, glycopeptides could be efficiently and selectively enriched with good reproducibility based on hydrophilic interaction chromatography (HILIC). Furthermore, the enrichment was specially achieved in weak alkaline environment, with 10 mM NH4HCO3 as the elution buffer, compatible with deglycosylation conditions. Therefore, the glycopeptides could be on-line deglycosylated with high efficiency and throughput by directly coupling the PNGase F functionalized monolithic column with the enrichment column during elution without the requirement of buffer exchange and pH adjustment. By such a method, within only 70-min pretreatment, 196 N-linked glycopeptides, corresponding to 122 glycoproteins, could be identified from 5 μg of human plasma with 14 high-abundant proteins removed, and the N-linked glycopeptides occupied 81% of all identified peptides, achieving to the best of our knowledge, the highest selectivity of HILIC-based methods. All the results demonstrated the high efficiency, selectivity and throughput of our proposed strategy for the large scale glycoproteome analysis.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Cystamine dihydrochloride, 96%
Sigma-Aldrich
Cystamine dihydrochloride, BioXtra
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
DL-Cysteine, technical grade
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)