Merck
  • An in situ gelling liquid crystalline system based on monoglycerides and polyethylenimine for local delivery of siRNAs.
An in situ gelling liquid crystalline system based on monoglycerides and polyethylenimine for local delivery of siRNAs.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-04-29)
Lívia Neves Borgheti-Cardoso, Lívia Vieira Depieri, Sander A A Kooijmans, Henrique Diniz, Ricardo Alexandre Junqueira Calzzani, Fabiana Testa Moura de Carvalho Vicentini, Roy van der Meel, Márcia Carvalho de Abreu Fantini, Mamie Mizusaki Iyomasa, Raymond M Schiffelers, Maria Vitória Lopes Badra Bentley
ABSTRACT
The development of delivery systems able to complex and release siRNA into the cytosol is essential for therapeutic use of siRNA. Among the delivery systems, local delivery has advantages over systemic administration. In this study, we developed and characterized non-viral carriers to deliver siRNA locally, based on polyethylenimine (PEI) as gene carrier, and a self-assembling drug delivery system that forms a gel in situ. Liquid crystalline formulations composed of monoglycerides (MO), PEI, propylene glycol (PG) and 0.1M Tris buffer pH 6.5 were developed and characterized by polarized light microscopy, Small Angle X-ray Scattering (SAXS), for their ability to form inverted type liquid crystalline phases (LC2) in contact with excess water, water absorption capacity, ability to complex with siRNA and siRNA release. In addition, gel formation in vivo was determined by subcutaneous injection of the formulations in mice. In water excess, precursor fluid formulations rapidly transformed into a viscous liquid crystalline phase. The presence of PEI influences the liquid crystalline structure of the LC2 formed and was crucial for complexing siRNA. The siRNA was released from the crystalline phase complexed with PEI. The release rate was dependent on the rate of water uptake. The formulation containing MO/PEI/PG/Tris buffer at 7.85:0.65:76.5:15 (w/w/w/w) complexed with 10 μM of siRNA, characterized as a mixture of cubic phase (diamond-type) and inverted hexagonal phase (after contact with excess water), showed sustained release for 7 days in vitro. In mice, in situ gel formation occurred after subcutaneous injection of the formulations, and the gels were degraded in 30 days. Initially a mild inflammatory process occurred in the tissue surrounding the gel; but after 14 days the tissue appeared normal. Taken together, this work demonstrates the rational development of an in situ gelling formulation for local release of siRNA.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
SAFC
Tromethamine