Merck
  • Mutation analysis of PVRL1 in patients with non-syndromic cleft of the lip and/or palate in Guangdong.
Mutation analysis of PVRL1 in patients with non-syndromic cleft of the lip and/or palate in Guangdong.
Genetics and molecular research : GMR (2015-05-13)
S Y Shu, M J Zhang, H Q Cheng, S J Tang, W L Chen, S R Wu, Y Lin, Q S Chen
ABSTRACT
Non-syndromic cleft of the lip and/or palate (NSCLP) is a very common birth defect; the poliovirus receptor-like 1 gene (PVRL1) has been identified as a genetic risk factor for NSCLP in patients from Norway, the Philippines, and South America. Given the considerable variation in allele frequencies across these geographical regions, this study explored the relationship between NSCLP and mutations of PVRL1 in patients from Guangdong, China. We recruited 171 NSCLP patients and 100 volunteers, and divided our samples into 2 groups: a sequencing group and a mass spectrometry group. In the sequencing group, we screened for mutations in exons 2 and 5 of PVRL1 by polymerase chain reaction and direct sequencing in 71 NSCLP patients and 100 volunteers. In the mass spectrometry group, we screened for amino acid mutations in α-spliced transcript codons 112, 131, and 395, and in the β-spliced transcript codon 1082 using matrix-assisted laser desorption/ionization time-of-flight mass spectrometry analysis in 100 NSCLP patients and 100 volunteers. No mutations were detected in either PVRL1 exons 2 or 5 in the 71 NSCLP patients and 100 volunteers, nor did we find mutations of α-spliced transcript codons 112, 131, 395 and the β-spliced transcript codon 1082 in any of the 100 NSCLP patients and 100 volunteers. Thus, mutations in exons 2 and 5 of PVRL1, and T334A, A391T, G1183A in the α-spliced transcript, and G1082T in the β-spliced transcript do not participate in the development of NSCLP in patients from Guangdong.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)