Merck
  • Chemo-Enzymatic Synthesis of Linear and Branched Cationic Peptides: Evaluation as Gene Carriers.
Chemo-Enzymatic Synthesis of Linear and Branched Cationic Peptides: Evaluation as Gene Carriers.
Macromolecular bioscience (2015-04-02)
Jose Manuel Ageitos, Jo-Ann Chuah, Keiji Numata
ABSTRACT
Cationic peptides such as poly(l-lysine) and poly(l-arginine) are important tools for gene delivery since they can efficiently condense DNA. It is difficult to produce cationic peptides by recombinant bacterial expression, and its chemical synthesis requires several steps of protection/deprotection and toxic agents. Chemo-enzymatic synthesis of peptides is a clean chemistry technique that allows fast production under mild conditions. With the aim to simplify the production of cationic peptides, the present work develops an enzymatic reaction which enables the synthesis of linear cationic peptides and, through terminal functionalization with tris(2-aminoethyl)amine, of branched cationic peptide conjugates, which show improved DNA complex formation. Cytotoxicity and transfection efficiency of all the chemo-enzymatically synthesized cationic peptides are evaluated for their novel use as gene delivery agents. Synthesized peptides exhibit transfection efficiencies comparable to previously reported monodisperse peptides. Chemo-enzymatic synthesis opens the door for efficient production of cationic peptides for their use as gene delivery carriers.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Tris(2-aminoethyl)amine, 96%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC