Merck
  • Biomimetic fiber assembled gradient hydrogel to engineer glycosaminoglycan enriched and mineralized cartilage: An in vitro study.
Biomimetic fiber assembled gradient hydrogel to engineer glycosaminoglycan enriched and mineralized cartilage: An in vitro study.
Journal of biomedical materials research. Part A (2015-05-28)
Neethu Mohan, Jijo Wilson, Dexy Joseph, Dhanesh Vaikkath, Prabha D Nair
ABSTRACT
The study investigated the potential of electrospun fiber assembled hydrogel, with physical gradients of chondroitin sulfate (CS) and sol-gel-derived bioactive glass (BG), to engineer hyaline and mineralized cartilage in a single 3D system. Electrospun poly(caprolactone) (PCL) fibers incorporated with 0.1% w/w of CS (CSL) and 0.5% w/w of CS (CSH), 2.4% w/w of BG (BGL) and 12.5% w/w of BG (BGH) were fabricated. The CS showed a sustained release up to 3 days from CSL and 14 days from CSH fibers. Chondrocytes secreted hyaline like matrix with higher sulfated glycosaminoglycans (sGAG), collagen type II and aggrecan on CSL and CSH fibers. Mineralization was observed on BGL and BGH fibers when incubated in simulated body fluid for 14 days. Chondrocytes cultured on these fibers secreted a mineralized matrix that consisted of sGAG, hypertrophic proteins, collagen type X, and osteocalcin. The CS and BG incorporated PCL fiber mats were assembled in an agarose-gelatin hydrogel to generate a 3D hybrid scaffold. The signals in the fibers diffused and generated continuous opposing gradients of CS (chondrogenic signal) and BG (mineralization) in the hydrogel. The chondrocytes were encapsulated in hybrid scaffolds; live dead assay at 48 h showed viable cells. Cells maintained their phenotype and secreted specific extracellular matrix (ECM) in response to signals within the hydrogel. Continuous opposing gradients of sGAG enriched and mineralized ECM were observed surrounding each cell clusters on gradient hydrogel after 14 days of culture in response to the physical gradients of raw materials CS and BG. A construct with gradient mineralization might accelerate integration to subchondral bone during in vivo regeneration.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Safranin O, suitable for microscopy