Merck
  • Sporosarcina pasteurii use in extreme alkaline conditions for recycling solid industrial wastes.
Sporosarcina pasteurii use in extreme alkaline conditions for recycling solid industrial wastes.
Journal of biotechnology (2015-09-17)
Oana A Cuzman, Silvia Rescic, Katharina Richter, Linda Wittig, Piero Tiano
ABSTRACT
The ureolytic bacteria are one of the most efficient organisms able to produce high amounts of carbonate that easily react with the free calcium ions from the environment. Sporosarcina pasteurii, a robust microbe in alkaline environments, was tested in this work for its potential use in an eco-cementation process that involves the biomediated calcite precipitation (BCP). Bacterial behavior in extreme alkaline environment (pH values of 9-13) was tested in controlled laboratory conditions and in the presence of solid industry wastes, such as Cement Kiln Dust (CKD) and Lime Kiln Dust (LKD), by evaluating the enzymatic activity and the calcite precipitation capacity. Grain consolidation potential of S. pasteurii was tested for one type of CKD mixed with ground granulated blast-furnace slag (GGBS), with possible bioclogging and biocementation applications. The results revealed the formation of stable biocalcite in the presence of CKD, with a performance depending on the pH-value and free calcium ion content. The BCP induced by S. pasteurii and the recycling of solid wastes, such as CKD with high lime content, is a promising way for different bioclogging and biocementation applications, with benefits in construction costs and reduction of environmental pollution.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%