Merck
  • TTK activates Akt and promotes proliferation and migration of hepatocellular carcinoma cells.
TTK activates Akt and promotes proliferation and migration of hepatocellular carcinoma cells.
Oncotarget (2015-09-30)
Xing Liu, Weijia Liao, Qing Yuan, Ying Ou, Jian Huang
ABSTRACT
Hepatocellular carcinoma (HCC) is one of the most malignant cancers with poor clinical outcome. The protein kinase human monopolar spindle 1 (hMps1/TTK) gene expression is significantly increased in HCCs. However, its contributions to hepatocarcinogenesis remain unclear. In this study, we found that TTK was overexpressed in 77.63% (118/152) HCC specimens. Elevated TTK expression positively correlated with large tumor size and presence of the portal vein tumor thrombus (PVTT). Demethylation in its promoter increased TTK expression in HCC. In vitro assays revealed that TTK not only promoted cell proliferation and anchorage-independent growth, but also cell migration. Subsequent investigations revealed that TTK activated Akt/mTOR pathway in a p53 dependent manner. We also found that TTK specific kinase inhibitor AZ3146 could decrease HCC cell growth. In conclusion, TTK contributes to HCC tumorigenesis via promoting cell proliferation and migration. It may serve as a novel biomarker and a potential target in HCC cancer therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ttk
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
MISSION® esiRNA, targeting human TTBK2
Sigma-Aldrich
MISSION® esiRNA, targeting human TTK