Merck
  • Characterization of biomimetic calcium phosphate labeled with fluorescent dextran for quantification of osteoclastic activity.
Characterization of biomimetic calcium phosphate labeled with fluorescent dextran for quantification of osteoclastic activity.
Acta biomaterialia (2015-04-02)
Salwa M Maria, Christiane Prukner, Zeeshan Sheikh, Frank A Müller, Svetlana V Komarova, Jake E Barralet
ABSTRACT
Bone resorbing osteoclasts represent an important therapeutic target for diseases associated with bone and joint destruction, such as rheumatoid arthritis, periodontitis, and osteoporosis. The quantification of osteoclast resorptive activity in vitro is widely used for screening new anti-resorptive medications. The aim of this paper was to develop a simplified semi-automated method for the quantification of osteoclastic resorption using fluorescently labeled biomimetic mineral layers which can replace time intensive, often subjective and clearly non-sustainable use of translucent slices of tusks from vulnerable or endangered species such as the elephant. Osteoclasts were formed from RAW 264.7 mouse monocyte cell line using the pro-resorptive cytokine receptor activator of nuclear factor kappa-B ligand (RANKL). We confirmed that fluorescent labeling did not interfere with the biomimetic features of hydroxyapatite, and developed an automated method for quantifying osteoclastic resorption. Correlation between our assay and traditional manual measurement techniques was found to be very strong (R(2)=0.99). In addition, we modified the technique to provide depth and volume data of the resorption pits by confocal imaging at defined depths. Thus, our method allows automatic quantification of total osteoclastic resorption as well as additional data not obtainable by the current tusk slice technique offering a better alternative for high throughput screening of potential antiresorptives.
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals