Merck
  • Study on clarithromycin lactobionate based dual selector systems for the enantioseparation of basic drugs in capillary electrophoresis.
Study on clarithromycin lactobionate based dual selector systems for the enantioseparation of basic drugs in capillary electrophoresis.
Journal of separation science (2015-06-23)
Tao Yu, Yingxiang Du, Jiaquan Chen, Guangfu Xu, Ke Yang, Qi Zhang, Jinjing Zhang, Shuaijing Du, Zijie Feng, Yanjie Zhang
ABSTRACT
In this paper, the use of clarithromycin lactobionate, a kind of antibiotic chiral selector, in combination with four neutral cyclodextrin derivatives (glucose-β-cyclodextrin, hydroxyethyl-β-cyclodextrin, methyl-β-cyclodextrin and hydroxypropyl-β-cyclodextrin) was reported for the first time. As a result, these dual systems gave much better resolution of nefopam (the Rs increased to 3.58, 2.72, 1.49 and 1.42, respectively) compared to the single systems. The effects of buffer pH and selector concentration on the separation of nefopam were also investigated. Additionally, some other basic drugs including metoprolol, atenolol, propranolol, bisoprolol, esmolol and ritodrine were tested for the investigation and evaluation of the enantiorecognition capability of the four dual systems. As expected, the synergistic effect was observed in four systems. Different results of these dual systems were also summarized.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Sodium tetraborate decahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%