Merck
  • Large-scale preparation of graphene by high temperature insertion of hydrogen into graphite.
Large-scale preparation of graphene by high temperature insertion of hydrogen into graphite.
Nanoscale (2015-06-09)
Ali Reza Kamali, Derek J Fray
ABSTRACT
Experimental evidence for high temperature diffusion of hydrogen into the interlayer space of graphite is provided. This process is discussed as a possible method for the rapid production of high-quality, inexpensive graphene in large quantities, which could lead to the widespread application of graphene. It was found that hydrogen cations, dissolved in molten LiCl, can be discharged on cathodically polarized graphite rods, which then intercalate into the graphite structure, leading to the peeling of graphite to produce graphene. The graphene nanosheets produced displayed a single-crystalline structure with a lateral size of several hundred nanometers and a high degree of crystallinity and thermal stability. The method introduced could be scaled up to produce industrial quantities of high-quality graphene.
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%