Merck
  • A single exposure of sediment sulphate-reducing bacteria to oxytetracycline concentrations relevant to aquaculture enduringly disturbed their activity, abundance and community structure.
A single exposure of sediment sulphate-reducing bacteria to oxytetracycline concentrations relevant to aquaculture enduringly disturbed their activity, abundance and community structure.
Journal of applied microbiology (2015-05-15)
M L Fernández, F Granados-Chinchilla, C Rodríguez
ABSTRACT
Although feed medicated with antibiotics is widely used in animal production to prevent and treat bacterial infections, the effect of these drugs on nontarget anaerobic bacteria is unknown. We aimed to clarify whether a single exposure of sulphate-reducing bacteria (SRB) from a tilapia pond to oxytetracycline (OTC) concentrations relevant to aquaculture impacts their function, abundance and community structure. To demonstrate changes in SO4(2-) content, SRB abundance, dsrB copy number and SRB diversity, sediment mesocosms were spiked with 5, 25, 50 and 75 mg OTC kg(-1) and examined for 30 days by means of ion chromatography, qPCR, cultivation and fluorescent in situ hybridization (FISH). On day 3, we measured higher SO4(2-) concentrations (ca. two-fold) and a reduction in dsrB copy numbers of approximately 50% in the treatments compared to the controls. After 30 days, a subtle yet measurable enrichment of bacteria from the order Desulfovibrionales occurred in mesocosms receiving ≥ 50 mg OTC kg(-1), notwithstanding that SRB counts decreased two orders of magnitude. OTC was dynamically and reversibly converted into 4-epioxytetracycline and other related compounds in a dose-dependent manner during the experiment. A single exposure to rather high OTC concentrations triggered functional and structural changes in a SRB community that manifested quickly and persisted for a month. This study improves our limited knowledge on the ecotoxicology of antibiotics in anaerobic environments.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)