Merck
  • Using a decellularized splenic matrix as a 3D scaffold for hepatocyte cultivation in vitro: a preliminary trial.
Using a decellularized splenic matrix as a 3D scaffold for hepatocyte cultivation in vitro: a preliminary trial.
Biomedical materials (Bristol, England) (2015-08-21)
Xing-Long Zheng, Jun-Xi Xiang, Wan-Quan Wu, Bo Wang, Wen-Yan Liu, Rui Gao, Ding-Hui Dong, Yi Lv
ABSTRACT
Using a decellularized liver matrix (DLM) to reengineer liver tissue is a promising therapy for end-stage liver disease. However, the limited supply of donor organs still hampers its potential clinical application, while a xenogenic decellularized matrix may bring a risk of zoonosis and immunological rejection. Therefore, an appropriate alternative scaffold is needed. In this research, we established a decellularized splenic matrix (DSM) in a rodent model, which preserved the 3D ultrastructure, the components of the extracellular matrix (ECM) and the native vascular network. The DSM and DLM had similar components of ECM, and similar mechanical properties. Hepatocytes were seeded to the DSM and DLM for dynamic culturing up to 6 d, and distributed both in decellularized sinusoidal spaces and around the vessels. The TUNEL-positive cell percentage in a dynamic culturing decellularized splenic matrix (dDSM) was 10.7%  ±  3.6% at 3d and 25.8%  ±  5.6% at 5d, although 14.2%  ±  4.5% and 24.8%  ±  2.9%, respectively, in a dynamic culturing decellularized liver matrix (dDLM) at the same time point (p  >  0.05). Primary hepatocytes in the dDSM and dDLM expressed albumin, G6pc and Ugt1a1. The gene expression of Cyp2b1, Cyp1a2 and HNF1α in the gene transcription level revealed hepatocytes had lower gene expression levels in the dDSM compared with the dDLM at 3d, but better than those in a sandwich culture. The cumulative albumin production at 6 d of culture was 80.7   ±   9.6 μg per million cells in the dDSM and 89.6   ±   4.6 μg per million cells in the dDLM (p  >  0.05). In summary, the DSM is a promising 3D scaffold for hepatocyte cultivation in vitro.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%