Merck
  • Randomised controlled trial of the effect of long-term selenium supplementation on plasma cholesterol in an elderly Danish population.
Randomised controlled trial of the effect of long-term selenium supplementation on plasma cholesterol in an elderly Danish population.
The British journal of nutrition (2015-10-01)
Frederik Cold, Kristian H Winther, Roberto Pastor-Barriuso, Margaret P Rayman, Eliseo Guallar, Mads Nybo, Bruce A Griffin, Saverio Stranges, Søren Cold
ABSTRACT
Although cross-sectional studies have shown a positive association between Se and cholesterol concentrations, a recent randomised controlled trial in 501 elderly UK individuals of relatively low-Se status found that Se supplementation for 6 months lowered total plasma cholesterol. The Danish PRECISE (PREvention of Cancer by Intervention with Selenium) pilot study (ClinicalTrials.gov ID: NCT01819649) was a 5-year randomised, double-blinded, placebo-controlled trial with four groups (allocation ratio 1:1:1:1). Men and women aged 60-74 years (n 491) were randomised to 100 (n 124), 200 (n 122) or 300 (n 119) μg Se-enriched yeast or matching placebo-yeast tablets (n 126) daily for 5 years. A total of 468 participants continued the study for 6 months and 361 participants, equally distributed across treatment groups, continued for 5 years. Plasma samples were analysed for total and HDL-cholesterol and for total Se concentrations at baseline, 6 months and 5 years. The effect of different doses of Se supplementation on plasma lipid and Se concentrations was estimated by using linear mixed models. Plasma Se concentration increased significantly and dose-dependently in the intervention groups after 6 months and 5 years. Total cholesterol decreased significantly both in the intervention groups and in the placebo group after 6 months and 5 years, with small and nonsignificant differences in changes in plasma concentration of total cholesterol, HDL-cholesterol, non-HDL-cholesterol and total:HDL-cholesterol ratio between intervention and placebo groups. The effect of long-term supplementation with Se on plasma cholesterol concentrations or its sub-fractions did not differ significantly from placebo in this elderly population.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.