Merck
  • Clinical survival of indirect, anterior 3-unit surface-retained fibre-reinforced composite fixed dental prosthesis: Up to 7.5-years follow-up.
Clinical survival of indirect, anterior 3-unit surface-retained fibre-reinforced composite fixed dental prosthesis: Up to 7.5-years follow-up.
Journal of dentistry (2015-04-29)
Ovul Kumbuloglu, Mutlu Özcan
ABSTRACT
This prospective clinical study evaluated the performance of indirect, anterior, surface-retained, fibre-reinforced-composite restorations (ISFRCR). Between June-2003 and January-2011, a total of 134 patients (83 females, 51 males, 16-68 years old) received 175 ISFRCRs (local ethical registration number: 14/9/4). All restorations were made indirectly on a plaster model using unidirectional E-glass fibres (everStick C&B, StickTech) in combination with a laboratory resin composite (Dialogue, Schütz Dental) and cemented according to the instructions of 4 resin cements [(RelyX ARC, 3M-ESPE, n=61), Bifix DC, VOCO, n=45), Variolink II (Ivoclar Vivadent, n=32) and Multilink (Ivoclar Vivadent, n=37)]. After baseline recordings, patients were followed at 6 months and thereafter annually up to 7.5 years. The evaluation protocol involved technical (chipping, debonding or fracture of tooth/restoration) and biological failures (caries). Mean observation period was 58 months. Altogether, 13 failures were observed [survival rate: 97.7%] (Kaplan-Meier). One catastrophic fracture [(cement: RelyX ARC), eight partial debonding (cement: Bifix DC (5), Multilink (1), RelyX ARC (1), Variolink II (1)] and four delaminations of veneering composite [(cement: Bifix DC (2), RelyX ARC (1), Multilink (1)] were observed. Except one replacement, all defective restorations were repaired or recemented. Annual failure rate of ISFRCRs was 1.73%. The survival rates with the four resin cements did not show significant differences (RelyX ARC: 98.3%; Bifix DC: 93.5%; Variolink 2: 100%; Multilink: 100%) (p=0.114). Secondary caries did not occur in any of the teeth. The 3-unit anterior indirect surface-retained resin-bonded FRC FDPs showed similar clinical survival rate when cemented with the resin cements tested. Experienced failures in general were due to debonding of the restoration or delamination of the veneering composite. 3-unit surface retained resin-bonded FRC FDPs could be considered minimal invasive and cost-effective alternatives to conventional tooth- or implant-borne FDPs. Failures were mainly repairable in the form of chipping or debonding depending on the resin cement type.
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Ytterbium(III) fluoride, anhydrous, powder, 99.98% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis