Merck
  • Effect of concentrate feeder design on performance, eating and animal behavior, welfare, ruminal health, and carcass quality in Holstein bulls fed high-concentrate diets.
Effect of concentrate feeder design on performance, eating and animal behavior, welfare, ruminal health, and carcass quality in Holstein bulls fed high-concentrate diets.
Journal of animal science (2015-06-27)
M Verdú, A Bach, M Devant
ABSTRACT
A total of 240 Holstein bulls (121 ± 2.0 kg initial BW; 99 ± 1.0 d of age), from 2 consecutive fattening cycles, were randomly allocated in 1 of 6 pens and assigned to 1 of the 3 treatments consisting of different concentrate feeder designs: a control feeder with 4 feeding spaces (CF), a feeder with less concentrate capacity (CFL), and a single-space feeder with lateral protections (SF). Each pen had a straw feeder and a drinker. All animals were fed a high-concentrate diet for ad libitum intake. Concentrate consumption was recorded daily using a computerized feeder, straw consumption was recorded weekly, and BW was recorded every 14 d. Animal behavior was registered on d 1, 3, 5, 8, and 14 and every 28 d by scan sampling. Eating behavior at concentrate feeders was filmed on d 12, 125, and 206. On d 7, 120, and 204, samples of rumen contents were collected for measurement of pH and VFA and blood samples were obtained to analyze NEFA, haptoglobin, glucose, and insulin. Animals were slaughtered after 223 d, and HCW and lesions of the rumen wall and liver were recorded. The accumulative concentrate consumption per animal tended (P = 0.09) to be greater with CF than with CFL and SF. Also, CV of concentrate consumption was greater (P < 0.01) for SF than for CF or CFL. However, feeder design did not influence the other performance and carcass data. Also, no differences among treatments in rumen wall evaluation and liver abscesses were observed. At 7 and 204 d of study, SF bulls had greater (P < 0.05) rumen pH compared with CF and CFL bulls. On d 7, the acetate to propionate ratio from SF was greater (P < 0.05) than for CFL or CF. At d 7, NEFA of SF were greater (P < 0.05) compared with CF and CFL. Bulls fed with CF have the greatest (P < 0.01) concentrate disappearance velocity followed by bulls fed with CFL and finally by bulls fed with SF, and this was associated with different feeding behaviors. Bulls on SF spent more time (P < 0.05) eating straw and exhibited fewer (P < 0.05) displacements at concentrate feeder than CF and CFL bulls. The CFL bulls exhibited (P < 0.01) more attempted mounts and tended (P = 0.10) to exhibit more completed mounts than CF bulls. In conclusion, both alternative feeder designs (CFL and SF) are good strategies to reduce total concentrate consumption without impairing performance, rumen health, or animal welfare in Holstein bulls fed high-concentrate diets. However, at the beginning, there was evidence that animals fed using SF had problems with adaptation.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®