Merck
  • Synthesis and characterization of phosphodiester stationary bonded phases for liquid chromatography.
Synthesis and characterization of phosphodiester stationary bonded phases for liquid chromatography.
Talanta (2015-06-17)
Szymon Bocian, Bogusław Buszewski
ABSTRACT
The new type of stationary bonded phases for liquid chromatography with immobilized artificial membrane properties was synthesized. Based on the modification of diol-bonded silica gel the phosphodiester stationary phases were obtained. The structures of synthesized material were confirmed by different physico-chemical techniques such as elemental analysis, infrared spectroscopy (FTIR), (13)C CP/MAS NMR and in reversed phase and hydrophilic interaction liquid chromatography, where hydrophobic and polar compounds were sufficiently separated. To present the influence of phosphate group on the retention properties, obtained material were compared with Diol-Ester C12 stationary phase.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
1-Decanol, 98%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
1-Octadecanol, 95%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
1-Decanol, ≥98%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Rhein
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%