Merck
  • Stomatitis Associated With Use of mTOR Inhibitors: Implications for Patients With Invasive Breast Cancer.
Stomatitis Associated With Use of mTOR Inhibitors: Implications for Patients With Invasive Breast Cancer.
Clinical journal of oncology nursing (2015-07-26)
Josephine Divers, Joyce O'Shaughnessy
ABSTRACT
The mammalian target of rapamycin (mTOR) inhibitor everolimus is approved (in combination with exemestane) for the treatment of postmenopausal women with advanced hormone receptor-positive, human epidermal growth factor receptor 2-negative breast cancer resistant to endocrine therapy. Stomatitis is among the most frequently reported dose-limiting adverse events associated with everolimus use, often requiring treatment interruption or dose reduction. This article aims to educate nurses on the identification and management of stomatitis associated with mTOR inhibitors in hormone receptor-positive advanced breast cancer and to assist nurses with additional management techniques to improve patient outcomes. An evaluation of the literature highlighting the incidence, identification, and management of stomatitis in cancer was performed with a particular focus on breast cancer. In addition, the experiences of the authors' cancer center on managing stomatitis are described. A growing body of clinical evidence shows the benefits of adding steroid-based mouth rinses to the treatment plan. Clinical experience provides additional insight into stomatitis preventive and management strategies for patients with breast cancer receiving treatment with everolimus.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, natural, 97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Triamcinolone
Pricing and availability is not currently available.