Merck
  • Assessment of alkaline cholesterol oxidase purified from Rhodococcus sp. PKPD-CL for its halo tolerance, detergent and organic solvent stability.
Assessment of alkaline cholesterol oxidase purified from Rhodococcus sp. PKPD-CL for its halo tolerance, detergent and organic solvent stability.
Protein expression and purification (2015-08-16)
Pramod J Kasabe, Geetanjali T Mali, Padma B Dandge
ABSTRACT
The novel bacterium, Rhodococcus sp. PKPD-CL was isolated and identified from the 'Chilika Lake' located at Odisha state of India, which is a largest brackish water habitat in Asia. Rhodococcus sp. PKPD-CL produces extracellular halo tolerant, detergent and organic solvent stable alkaline cholesterol oxidase. It has apparent molecular weight of 60 kDa and was purified 59 fold by using 60% saturated ammonium sulfate fractionation, anion exchange followed by size exclusion chromatographic techniques with 37% recovery. It showed substrate specificity for 3β-hydroxysteroids with Km of 1.1 × 10(-4)M for cholesterol. The pH, 8.0 and the temperature, 37 °C were required for its optimum activity. Enzyme is considerably stable at pH 6.0-8.5 and temperature up to 50 °C. At 4 and 30 °C it maintained its 100% activity up to 60 days. The isoelectric point of the enzyme was 9.5. It showed 80% residual activity with 20% NaCl (3.42 M) and 83% relative activity with 12% NaCl (2.05 M) concentration. The metal ions like Zn(2+), Cu(2+), Ag+, Fe(3+), Ba(2+) inhibited the enzyme activity >60% while Hg(2+) served a potent inhibitor whereas Mg(2+) found to be a good enhancer for it. The enzyme was stable in presence of chemical reagents (NaN3, EDTA), detergents (Tween-80, Tween-20, Triton X-100, sodium cholate) and various organic solvents (isopropanol, ethanol, benzene, chloroform, methanol, toluene, ethyl acetate, butanol and dimethylsulfoxide). Such a multi stress tolerant and versatile enzyme produced by Rhodococcus sp. PKPD-CL may serve as a good choice for industrial applications.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Cholesteryl linoleate, ≥98% (HPLC; detection at 205 nm)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Phenol, natural, 97%, FG