Merck
Functional synergism of Human Defensin 5 and Human Defensin 6.
Biochemical and biophysical research communications (2015-10-18)
Aiping Zhao, Wuyuan Lu, Erik de Leeuw
ABSTRACT
The gut epithelium is critically involved in maintaining intestinal immune homeostasis. Acting as a physical barrier, it separates the intestinal microflora from cells of the immune system. In addition to its barrier function, the intestinal epithelium expresses defensins, natural, endogenous antimicrobial peptides. In humans, specialized epithelial cells, termed Paneth cells, located primarily in the small intestine express two defensins, Human Defensin-5 (HD-5) and Human Defensin-6 (HD-6). Previously, we have shown that HD-5 potently kills bacteria and induces secretion of interleukin-8 by intestinal epithelial cells. We show that HD-6 specifically and synergistically enhances the HD-5-induced IL-8 secretion, but does not alter its anti-bacterial activity. Further, we find that HD-5 decreases the trans-epithelial electrical resistance of intestinal epithelial cells and that HD-6 negates this effect of HD-5.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
MSTFA Reagent, ampule of 10 × 1.2 mL, analytical standard, Cerilliant®