Merck
  • Activities of genes controlling sphingolipid metabolism in human fibroblasts treated with flavonoids.
Activities of genes controlling sphingolipid metabolism in human fibroblasts treated with flavonoids.
Metabolic brain disease (2015-07-26)
Marta Moskot, Joanna Jakóbkiewicz-Banecka, Elwira Smolińska, Bogdan Banecki, Grzegorz Węgrzyn, Magdalena Gabig-Cimińska
ABSTRACT
Natural flavonoids such as genistein, kaempferol and daidzein were previously found to be able to reduce efficiency of glycosaminoglycan synthesis in cells of patients suffering from mucopolysaccharidoses, inherited metabolic diseases with often brain disease symptoms. This feature was employed to test these compounds as potential drugs for treatment other neuronopathic lysosomal storage disorders, in which errors in sphingolipid metabolism occur. In this report, on the basis of DNA microarray analyses and quantitative real time PCR experiments, we present evidence that these compounds modify expression of genes coding for enzymes required for metabolism of sphingolipids in human dermal fibroblasts (HDFa). Expression of several genes involved in sphingolipid synthesis was impaired by tested flavonoids. Therefore, it is tempting to speculate that they may be considered as potential drugs in treatment of LSD, in which accumulation of sphingolipids, especially glycosphingolipids, occurs. Nevertheless, further studies on more advances models are required to test this hypothesis and to assess a therapeutic potential for flavonoids in this group of metabolic brain diseases.
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%