Merck
  • The Occurrence of Propyl Lactate in Chinese Baijius (Chinese Liquors) Detected by Direct Injection Coupled with Gas Chromatography-Mass Spectrometry.
The Occurrence of Propyl Lactate in Chinese Baijius (Chinese Liquors) Detected by Direct Injection Coupled with Gas Chromatography-Mass Spectrometry.
Molecules (Basel, Switzerland) (2015-10-23)
Jihong Wu, Yang Zheng, Baoguo Sun, Xiaotao Sun, Jiyuan Sun, Fuping Zheng, Mingquan Huang
ABSTRACT
As one of the oldest distillates in the world, flavor compounds of Chinese Baijiu (Chinese liquor) were extremely complex. Propyl lactate was firstly detected by direct injection and gas chromatography-mass spectrometry (GC-MS) in 72 Chinese Baijius. The objectives were to detect the contents of propyl lactate and evaluate its contribution to the aroma of Chinese Baijiu based on odor activity values (OAVs). The levels of propyl lactate in these distillates were determined by internal standard method and selective ion monitoring (SIM), which ranged from 0.050 to 1.900 mg∙L(-1) under investigation. Its detection threshold was determined by Three-Alternative Forced-Choice (3-AFC) and curve fitting (CF), which was 0.740 mg∙L(-1) in 38% ethanol solution. The contribution of propyl lactate on the aroma of these distillate drinks was evaluated by their odor activity values (OAVs), which varied from 0.066 to 4.440. The OAVs of propyl lactate were found to exceed 1 in 13 Chinese Baijius, including 50° Jingzhi Guniang 5 years (4.440), 52° Jingzhi Guniang 10 years (3.024), Jingyanggang (2.568), Xianghe Ronghe Shaofang (2.313), and 1956 Laolang (1.431), which indicated that propyl lactate was one of odor-active components in these Chinese Baijius.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methyl DL-lactate, ≥97.0% (GC)