Merck
  • Design, development and in-vitro evaluation of diclofenac taste-masked orodispersible tablet formulations.
Design, development and in-vitro evaluation of diclofenac taste-masked orodispersible tablet formulations.
Drug development and industrial pharmacy (2014-02-06)
Marie Guhmann, Maren Preis, Frédéric Gerber, Norbert Pöllinger, Jörg Breitkreutz, Werner Weitschies
ABSTRACT
Fast onset of action is prerequisite for acute pain medication. A palatable orodispersible medicine of diclofenac providing rapid analgesic effect should improve patient compliance and treatment. In the present study, diclofenac taste-masked orodispersible tablets (ODTs) with fast release characteristics were developed. Different taste-masking approaches and formulation concepts were screened in vitro for candidate selection. Diclofenac was used as free acid. Five taste-masked microgranule formulations were prepared by wet granulation and/or coating processes, and compressed to ODTs. Citric acid (pH-modifying agent) and Eudragit® E PO (amino methacrylate copolymer) were used as taste-masking agents. Evaluation criteria were (i) disintegration time, (ii) processability and (iii) in-vitro dissolution profiles in simulated saliva (pH 7.4, 5 mL, 3 min) and compendial pH-change media (paddle, 50 rpm). The prototypes were compared to reference ODTs (without taste-masking). Most suitable ODT prototypes were selected and further evaluated for taste-masking efficiency using an electronic tongue. In simulated saliva, the drug was slower released from the prototypes (between 1.1% and 15.5%) than from reference ODTs (23.7%). Less dissolved particles are thus expected in vivo for taste perception. Two ODT prototypes showed fast and complete drug release in phosphate buffer. The formulation providing the most efficient taste-masking was selected guided by electronic tongue data. A novel palatable and fast acting diclofenac ODT formulation was successfully developed. Formulation design, development and in-vitro evaluation used in this study may serve as rational approach for manufacturing taste-masked orodispersible dosage forms.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis