Merck
Reactive Carbonyl Species Derived from Omega-3 and Omega-6 Fatty Acids.
Journal of agricultural and food chemistry (2015-07-08)
Yu Wang, Ping Cui
ABSTRACT
Inflammation-related reactive oxygen species (ROS) and reactive nitrogen species (RNS) are associated with the development of cancer. ROS and RNS can directly damage biomacromolecules such as proteins, DNA, and lipids. Lipid peroxidation, however, can result in reactive carbonyl species (RCS) that can also modify proteins and DNA. In contrast to an extensive literature on the modification of proteins and DNA from omega-6 fatty acids, there are few studies on RCS generation from other fatty acids, particularly omega-3 fatty acids, which are frequently consumed from the diet and diet supplements. Therefore, a comparison between omega-3 and omega-6 fatty acids has been conducted. LC-MS/MS analysis of carbonyl-dinitrophenylhydrazine (DNPH) standards yielded characteristic fragment ions. Autoxidation products of α-linolenic acid and linoleic acid were then derivatized with DNPH and analyzed by LC-MS/MS. The results showed that α-linolenic acid, an omega-3 fatty acid, generated more acrolein and crotonaldehyde than did linoleic acid, an omega-6 fatty acid. Omega-3 fatty acids might be easily degraded to smaller monoaldehydes or dicarbonyls. Omega-3 fatty acids have been considered as health improvement components for a long time. However, on the basis of the results presented here, use of omega-3 fatty acids should be re-evaluated in vivo for safety purposes.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
Sigma-Aldrich
Valeraldehyde, ≥97%, FG
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Valeraldehyde, 97%
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%