Merck
  • Synthesis of thiol derivatives of azobenzocrown ethers. The preliminary studies on recognition of alkali metal ions by gold nanoparticles functionalized with azobenzocrown and lipoic acid.
Synthesis of thiol derivatives of azobenzocrown ethers. The preliminary studies on recognition of alkali metal ions by gold nanoparticles functionalized with azobenzocrown and lipoic acid.
Journal of inclusion phenomena and macrocyclic chemistry (2015-11-10)
Elżbieta Luboch, Mirosław Szarmach, Anna Buczkowska, Ewa Wagner-Wysiecka, Magdalena Kania, Witold Danikiewicz
ABSTRACT
The article presents the synthesis of novel 13- and 16-membered azobenzocrown derivatives with peripheral thiol moieties and preliminary studies assessing their possible application in plasmonic sensors based on gold nanoparticles. The effect of the length of the chain connecting the macrocycle with the thiol group and the effect of the presence of the additional functional compound, i.e. lipoic acid, on the sensor response was analyzed. Colloidal gold nanoparticles modified with a 16-membered crown with a thiol group on oxyethylene (compound
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
(±)-α-Lipoic acid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Sigma-Aldrich
1,10-Dibromodecane, 97%
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
Tetraethylenepentamine, technical grade