Merck
  • Enzymatic hydrolysis of ovomucoid and the functional properties of its hydrolysates.
Enzymatic hydrolysis of ovomucoid and the functional properties of its hydrolysates.
Poultry science (2015-07-22)
E D N S Abeyrathne, H Y Lee, C Jo, J W Suh, D U Ahn
ABSTRACT
Ovomucoid is well known as a "trypsin inhibitor" and is considered to be the main food allergen in egg. However, the negative functions of ovomucoid can be eliminated if the protein is cut into small peptides. The objectives of this study were to hydrolyze ovomucoid using various enzyme combinations, and compare the functional properties of the hydrolysates. Purified ovomucoid was dissolved in distilled water (20 mg/mL) and treated with 1% of pepsin, α-chymotrypsin, papain, and alcalase, singly or in combinations. Sodium sodium dodecyl sulfate-polyacrylamide (SDS-PAGE) results of the hydrolysates indicated that pepsin (OMP), alcalase (OMAl), alcalase+trypsin (OMAlTr), and alcalase+papain (OMAlPa) treatments best hydrolyzed the ovomucoid, and the 4 treatments were selected to determine their functional characteristics. Among the 4 enzyme treatments, hydrolysate from OMAlTr showed the highest iron-chelating and antioxidant activities, while OMP showed higher ACE-inhibitory activity, but lower Fe-chelating activity than the other treatments. However, no difference in the copper-chelating activity among the treatments was found. MS/MS analysis identified numerous peptides from the hydrolysates of OMAlPa and OMAlTr, and majority of the peptides produced were <2 kDa. Pepsin treatment (OMP), however, hydrolyzed ovomucoid almost completely and produced only amino acid monomers, di- and tri-peptides. The ACE-inhibitory, antioxidant and iron-chelating activities of the enzyme hydrolysates were not consistent with the number and size of peptides in the hydrolysates, but we do not have information about the quantity of each peptide present in the hydrolysates at this point.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M