Merck
  • Quantitative evaluation of the lengths of homobifunctional protein cross-linking reagents used as molecular rulers.
Quantitative evaluation of the lengths of homobifunctional protein cross-linking reagents used as molecular rulers.
Protein science : a publication of the Protein Society (2001-06-23)
N S Green, E Reisler, K N Houk
ABSTRACT
Homobifunctional chemical cross-linking reagents are important tools for functional and structural characterization of proteins. Accurate measures of the lengths of these molecules currently are not available, despite their widespread use. Stochastic dynamics calculations now provide quantitative measures of the lengths, and length dispersions, of 32 widely used molecular rulers. Significant differences from published data have been found. See www.proteinscience.org
Sigma-Aldrich
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
Pricing and availability is not currently available.