Merck
  • Inhibition of Binding of β2-Glycoprotein 1 to Phosphatidylserine by Polymyxin B, a Lupus-Like Anticoagulant.
Inhibition of Binding of β2-Glycoprotein 1 to Phosphatidylserine by Polymyxin B, a Lupus-Like Anticoagulant.
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis (2013-11-28)
Boguslaw Uchman, Douglas A Triplett
ABSTRACT
Polymyxin B is a cationic peptide that inhibits phospholipid-dependent coagulation tests including activated partial thromboplastin time and to a lesser degree prothrombin time. Thrombin clotting time is insensitive to polymyxin B. β2-glycoprotein 1 (β2GP1) is a cofactor of antiphospholipid antibodies. Antiphospholipid autoantibodies also poses lupus anticoagulant activity through interactions with β2GP1. Using affinity chromatography, polymyxin B can effectively decrease the binding of β2GP1 to immobilize phosphatidylserine. Since then, anticoagulant effect of polymyxin B is most likely due to the binding to negatively charged phospholipids, preventing formation of coagulation complexes.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%