Merck
  • Targeting of mTORC2 may have advantages over selective targeting of mTORC1 in the treatment of malignant pheochromocytoma.
Targeting of mTORC2 may have advantages over selective targeting of mTORC1 in the treatment of malignant pheochromocytoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-02-11)
Xiaohua Zhang, Xianjin Wang, Tianyuan Xu, Shan Zhong, Zhoujun Shen
ABSTRACT
Recent studies have found that mammalian target of rapamycin complex 2 (mTORC2) is emerging as a potential therapeutic target in the treatment of many human cancers. However, the effects of targeting of mTORC2 on malignant pheochromocytomas (PCC) and paragangliomas (PGL) have not been reported. The aim of the study was to investigate the effects of targeting of mTORC2 on malignant PCC/PGL by comparing the inhibitory effects of targeting of mTORC2 with mTORC1 on pheochromocytoma PC12 cell in vitro and vivo. The expressions of regulatory-associated protein of mTOR (raptor) and rapamycin-insensitive companion of mTOR (rictor) were detected by immunohistochemistry in human tissues of malignant PCC. Targeting of mTORC1, mTORC2, and mTORC1/2 (mTORC1 and mTORC2) were performed by transfected with raptor, rictor, and mammalian target of rapamycin (mTOR) small interfering RNA (siRNA) in pheochromocytoma PC12 cell, respectively. MTT assay, apoptosis analysis, wound healing, and Transwell approach were performed. A tumor model in nude mice bearing PC12 cell xenografts, which were dosed with rapamycin or PP242, was established. The expression of raptor was frequently moderate positive, but the expression of rictor was frequently strong positive in malignant PCC. In vitro, although inhibition of mTORC1 was able to suppress PC12 cell proliferation, inhibition of mTORC2 more effectively suppressed cell proliferation. Inhibition of mTORC2 or mTORC1/2 more effectively prevented cell migration and invasion, and promoted cell apoptosis, while inhibition of mTORC1 only slightly prevented cell migration and invasion, and was not able to promoted apoptosis. Also, we found that mTOR downstream kinases were deregulated by targeting of mTORC2, but not mTORC1. In vivo, we found that PP242 was more potent than rapamycin in inhibiting tumor growth in tumor model. Our data suggest that targeting of mTORC2 may have advantages over selective targeting of mTORC1 in the treatment of malignant PCC/PGL. However, more clinical trials are needed to prove our findings.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human RORC
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rictor