Merck
  • Chronic lung infection by Pseudomonas aeruginosa biofilm is cured by L-Methionine in combination with antibiotic therapy.
Chronic lung infection by Pseudomonas aeruginosa biofilm is cured by L-Methionine in combination with antibiotic therapy.
Scientific reports (2015-11-03)
Divya Prakash Gnanadhas, Monalisha Elango, Akshay Datey, Dipshikha Chakravortty
ABSTRACT
Bacterial biofilms are associated with 80-90% of infections. Within the biofilm, bacteria are refractile to antibiotics, requiring concentrations >1,000 times the minimum inhibitory concentration. Proteins, carbohydrates and DNA are the major components of biofilm matrix. Pseudomonas aeruginosa (PA) biofilms, which are majorly associated with chronic lung infection, contain extracellular DNA (eDNA) as a major component. Herein, we report for the first time that L-Methionine (L-Met) at 0.5 μM inhibits Pseudomonas aeruginosa (PA) biofilm formation and disassembles established PA biofilm by inducing DNase expression. Four DNase genes (sbcB, endA, eddB and recJ) were highly up-regulated upon L-Met treatment along with increased DNase activity in the culture supernatant. Since eDNA plays a major role in establishing and maintaining the PA biofilm, DNase activity is effective in disrupting the biofilm. Upon treatment with L-Met, the otherwise recalcitrant PA biofilm now shows susceptibility to ciprofloxacin. This was reflected in vivo, in the murine chronic PA lung infection model. Mice treated with L-Met responded better to antibiotic treatment, leading to enhanced survival as compared to mice treated with ciprofloxacin alone. These results clearly demonstrate that L-Met can be used along with antibiotic as an effective therapeutic against chronic PA biofilm infection.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)