Merck
  • The functional dlt operon of Clostridium butyricum controls the D-alanylation of cell wall components and influences cell septation and vancomycin-induced lysis.
The functional dlt operon of Clostridium butyricum controls the D-alanylation of cell wall components and influences cell septation and vancomycin-induced lysis.
Anaerobe (2015-09-13)
Sandra Wydau-Dematteis, Mathilde Louis, Noël Zahr, René Lai-Kuen, Bruno Saubaméa, Marie-José Butel, Jean-Louis Pons
ABSTRACT
Clostridium butyricum is a Gram-positive bacterium involved in the development of necrotizing enterocolitis (NEC) in preterm infants. To colonize the digestive tract, components of the cell wall of C. butyricum must interact with the intestinal mucosa. The D-alanylation of cell wall components such as teichoic acids results in a net positive charge on the cell wall, which is important for many functions of Gram-positive bacteria. Notably, D-alanylation mediates resistance to antimicrobial peptides and antibiotics. Here, we show that the dlt operon of C. butyricum encodes the enzymes responsible for the D-alanylation of cell wall components and influences the surface properties of the cell wall. We show that the D-alanylation of cell wall components controls the septation of C. butyricum, which is an essential mechanism during vegetative growth. Furthermore, we find that D-alanylation is involved in the resistance of C. butyricum to some cationic antimicrobial peptides (CAMPs) and lysozyme. Finally, we show that the D-alanylation of cell wall components influences vancomycin-induced lysis.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%