Merck
  • Methyltetrahydrofolate vs Folic Acid Supplementation in Idiopathic Recurrent Miscarriage with Respect to Methylenetetrahydrofolate Reductase C677T and A1298C Polymorphisms: A Randomized Controlled Trial.
Methyltetrahydrofolate vs Folic Acid Supplementation in Idiopathic Recurrent Miscarriage with Respect to Methylenetetrahydrofolate Reductase C677T and A1298C Polymorphisms: A Randomized Controlled Trial.
PloS one (2015-12-03)
Azita Hekmatdoost, Farhad Vahid, Zahra Yari, Mohammadreza Sadeghi, Hassan Eini-Zinab, Niknam Lakpour, Soheila Arefi
ABSTRACT
To determine whether 5-methylenetetrahydrofolate (MTHF) is more effective than folic acid supplementation in treatment of recurrent abortion in different MTHFR gene C677T and A1298C polymorphisms. A randomized, double blind, placebo-controlled trial conducted April 2011-September 2014 in recurrent abortion clinics in Tehran, Iran. The participants were women with three or more idiopathic recurrent abortion, aged 20 to 45 years. Two hundred and twenty eligible women who consented to participate were randomly assigned to receive either folic acid or 5-MTHF according to the stratified blocked randomization by age and the number of previous abortions. Participants took daily 1 mg 5-methylentetrahydrofolate or 1 mg folic acid from at least 8 weeks before conception to the 20th week of the pregnancy. The primary outcome was ongoing pregnancy rate at 20th week of pregnancy, and the secondary outcomes were serum folate and homocysteine at the baseline, after 8 weeks, and at the gestational age of 4, 8, 12, and 20 weeks, MTHFR gene C677T and A1298C polymorphisms. There was no significant difference in abortion rate between two groups. Serum folate increased significantly in both groups over time; these changes were significantly higher in the group receiving 5-MTHF than the group receiving folic acid (value = 2.39, p<00.1) and the result was the same by considering the time (value = 1.24, p<0.01). Plasma tHcys decreased significantly in both groups over time; however these changes were not significantly different between the groups (value = 0.01, p = 0.47). The results do not support any beneficial effect of 5-MTHF vs. folate supplementation in women with recurrent abortion with any MTHFR C677T and/or A1298C polymorphism. ClinicalTrials.gov NCT01976676.
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (B12), analytical standard