Merck
  • A candidate reference measurement procedure for quantifying serum concentrations of 25-hydroxyvitamin D₃ and 25-hydroxyvitamin D₂ using isotope-dilution liquid chromatography-tandem mass spectrometry.
A candidate reference measurement procedure for quantifying serum concentrations of 25-hydroxyvitamin D₃ and 25-hydroxyvitamin D₂ using isotope-dilution liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-05-15)
Ekaterina M Mineva, Rosemary L Schleicher, Madhulika Chaudhary-Webb, Khin L Maw, Julianne C Botelho, Hubert W Vesper, Christine M Pfeiffer
ABSTRACT
The inaccuracy of routine serum 25-hydroxyvitamin D measurements hampers the interpretation of data in patient care and public health research. We developed and validated a candidate reference measurement procedure (RMP) for highly accurate quantitation of two clinically important 25-hydroxyvitamin D metabolites in serum, 25-hydroxyvitamin D2 [25(OH)D2] and 25-hydroxyvitamin D3 [25(OH)D3]. The two compounds of interest together with spiked deuterium-labeled internal standards [d 3-25(OH)D2 and d 6-25(OH)D3] were extracted from serum via liquid-liquid extraction. The featured isotope-dilution LC-MS/MS method used reversed-phase chromatography and atmospheric pressure chemical ionization in positive ion mode. A pentafluorophenylpropyl-packed UHPLC column together with isocratic elution allowed for complete baseline resolution of 25(OH)D2 and 25(OH)D3 from their structural C-3 isomers within 12 min. We evaluated method trueness, precision, potential interferences, matrix effects, limits of quantitation, and measurement uncertainty. Calibration materials were, or were traceable to, NIST Standard Reference Materials 2972. Within-day and total imprecision (CV) averaged 1.9 and 2.0% for 25(OH)D3, respectively, and 2.4 and 3.5% for 25(OH)D2, respectively. Mean trueness was 100.3% for 25(OH)D3 and 25(OH)D2. The limits of quantitation/limits of detection were 4.61/1.38 nmol/L for 25(OH)D3 and 1.46/0.13 nmol/L for 25(OH)D2. When we compared our RMP results to an established RMP using 40 serum samples, we found a nonsignificant mean bias of 0.2% for total 25(OH)D. This candidate RMP for 25(OH)D metabolites meets predefined method performance specifications (≤5% total CV and ≤1.7% bias) and provides sufficient sample throughput to meet the needs of the Centers for Disease Control and Prevention Vitamin D Standardization Certification Program. Graphical abstract Bias assessment using NIST standard reference materials. Legend CDC mean mass fractions (ng/g) ± U 95 (6 replicates per mean). NIST-certified mass fractions (ng/g) ± U 95 from the Certificates of Analysis.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.