Merck
  • Effect of various diets on the expression of phase-I drug-metabolizing enzymes in livers of mice.
Effect of various diets on the expression of phase-I drug-metabolizing enzymes in livers of mice.
Xenobiotica; the fate of foreign compounds in biological systems (2015-03-04)
Ying Guo, Julia Yue Cui, Hong Lu, Curtis D Klaassen
ABSTRACT
1. Previous studies have shown that diets can alter the metabolism of drugs; however, it is difficult to compare the effects of multiple diets on drug metabolism among different experimental settings. Phase-I-related genes play a major role in the biotransformation of pro-drugs and drugs. 2. In the current study, effects of nine diets on the mRNA expression of phase-I drug metabolizing enzymes in livers of mice were simultaneously investigated. Compared to the AIN-93M purified diet (control), 73 of the 132 critical phase-I drug-metabolizing genes were differentially regulated by at least one diet. Diet restriction produced the largest number of changed genes (51), followed by the atherogenic diet (27), high-fat diet (25), standard rodent chow (21), western diet (20), high-fructose diet (5), EFA deficient diet (3) and low n-3 FA diet (1). The mRNAs of the Fmo family changed most, followed by Cyp2b and 4a subfamilies, as well as Por (from 1121- to 21-fold increase of theses mRNAs). There were 59 genes not altered by any of these diets. 3. The present results may improve the interpretation of studies with mice and aid in determining effective and safe doses for individuals with different nutritional diets.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra