Merck
  • A Genome-Wide Scan Identifies Variants in NFIB Associated with Metastasis in Patients with Osteosarcoma.
A Genome-Wide Scan Identifies Variants in NFIB Associated with Metastasis in Patients with Osteosarcoma.
Cancer discovery (2015-06-19)
Lisa Mirabello, Roelof Koster, Branden S Moriarity, Logan G Spector, Paul S Meltzer, Joy Gary, Mitchell J Machiela, Nathan Pankratz, Orestis A Panagiotou, David Largaespada, Zhaoming Wang, Julie M Gastier-Foster, Richard Gorlick, Chand Khanna, Silvia Regina Caminada de Toledo, Antonio S Petrilli, Ana Patiño-Garcia, Luis Sierrasesúmaga, Fernando Lecanda, Irene L Andrulis, Jay S Wunder, Nalan Gokgoz, Massimo Serra, Claudia Hattinger, Piero Picci, Katia Scotlandi, Adrienne M Flanagan, Roberto Tirabosco, Maria Fernanda Amary, Dina Halai, Mandy L Ballinger, David M Thomas, Sean Davis, Donald A Barkauskas, Neyssa Marina, Lee Helman, George M Otto, Kelsie L Becklin, Natalie K Wolf, Madison T Weg, Margaret Tucker, Sholom Wacholder, Joseph F Fraumeni, Neil E Caporaso, Joseph F Boland, Belynda D Hicks, Aurelie Vogt, Laurie Burdett, Meredith Yeager, Robert N Hoover, Stephen J Chanock, Sharon A Savage
ABSTRACT
Metastasis is the leading cause of death in patients with osteosarcoma, the most common pediatric bone malignancy. We conducted a multistage genome-wide association study of osteosarcoma metastasis at diagnosis in 935 osteosarcoma patients to determine whether germline genetic variation contributes to risk of metastasis. We identified an SNP, rs7034162, in NFIB significantly associated with metastasis in European osteosarcoma cases, as well as in cases of African and Brazilian ancestry (meta-analysis of all cases: P = 1.2 × 10(-9); OR, 2.43; 95% confidence interval, 1.83-3.24). The risk allele was significantly associated with lowered NFIB expression, which led to increased osteosarcoma cell migration, proliferation, and colony formation. In addition, a transposon screen in mice identified a significant proportion of osteosarcomas harboring inactivating insertions in Nfib and with lowered NFIB expression. These data suggest that germline genetic variation at rs7034162 is important in osteosarcoma metastasis and that NFIB is an osteosarcoma metastasis susceptibility gene. Metastasis at diagnosis in osteosarcoma is the leading cause of death in these patients. Here we show data that are supportive for the NFIB locus as associated with metastatic potential in osteosarcoma.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Anti-NFIB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
o-Xylene, anhydrous, 97%